missing translation for 'onlineSavingsMsg'
Learn More

UPK1A Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Product Code. p-7211980 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16843855 20 μL 20µL
16833855 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 16843855

Brand: Proteintech 252751AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Component of the asymmetric unit membrane (AUM); a highly specialized biomembrane elaborated by terminally differentiated urothelial cells. May play an important role in normal bladder epithelial physiology, possibly in regulating membrane permeability of superficial umbrella cells or in stabilizing the apical membrane through AUM/cytoskeletal interactions.

Specifications

Antigen UPK1A
Applications Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene UPK1A
Gene Accession No. O00322, Q9D132
Gene Alias Tetraspanin 21, Tspan 21, TSPAN21, UP1A, UPIA, UPK1A, UPKA, uroplakin 1A, Uroplakin Ia
Gene Symbols Upk1a
Host Species Rabbit
Immunogen UPK1A Fusion Protein Ag17356
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 109637, 11045
Target Species Mouse, Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.