Learn More
Fisherbrand™ Universal V7 Replacement Lock Nut Pack
Product Code. 16206061 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Type:
Universal (V7) Replacement Lock Nut Pack
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
16206061 Universal (V7) Replacement Lock Nut Pack Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16206061
  • ONLINE EXCLUSIVE PRICE
    225.00 NOK
    193.44 NOK / Pack of 5
    Save 31.56 NOK
    14% Off
View all promotions
Estimated Shipment: 08-01-2027
Add to basket
Add to basket
For use with 10mm shafts.
  • Replacement set
  • For general laboratory use
Specifications
For Use With (Equipment) For 10mm shaft
Type Universal (V7) Replacement Lock Nut Pack