Learn More
Fisherbrand™ Universal Stirrer Guide Replacement Bearing Seal
Shop All Fisherbrand Products
Click to view available options
For Use With (Equipment):
For Shaft: 12 - 16 mm
Temperature (Metric):
-200°C to +280°C
Unit Size:
Each
Specifications
Specifications
| Description | Fisherbrand™ Universal Stirrer Guide Replacement Bearing Seal |
| For Use With (Equipment) | For Shaft: 12 - 16 mm |
| Quantity | 1 |
| Temperature (Metric) | -200°C to +280°C |