Learn More
Fisherbrand™ Universal pH Indicator Paper
Product Code. 11783554 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
pH Range:
0 to 10
1 to 14
5 to 9
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. pH Range Unit Size
11783554 0 to 10 Each
11703564 5 to 9 Each
11763554 1 to 14 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11783554
  • ONLINE EXCLUSIVE PRICE
    74.65 NOK
    67.03 NOK / Pack of 1
    Save 7.62 NOK
    10% Off
View all promotions
 Restricted Item
Estimated Shipment: 20-01-2027
Add to basket
Add to basket
Provides rapid and precise readings. Fisherbrand™ Universal pH Indicator Paper include a color scale printed on the cover for convenience.
  • Filter paper soaked with one or several colored indicators of pH
  • Economic range of pH 0 to 14
  • Available in a variety of formats to meet individual needs
  • Reference chart shows color changes in steps of pH2
Specifications
pH Range 0 to 10
Format Roll
Quantity 1 Roll
Type Grade 911
Length (Metric) 5 m
Width (Metric) 10 mm