Learn More
Fisherbrand™ Universal pH Indicator Paper
Product Code. 12345378 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
pH Range:
0.5 to 5
0.5 to 5.5
1 to 11
1 to 14
4 to 7
5 to 9
6.5 to 10
9 to 13
Format:
Roll
Tube
Length (Metric):
50 mm
5 m
Quantity:
1 Roll
10 Pack
4 Rolls
Unit Size:
Each
Pack of 4
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. pH Range Format Length (Metric) Quantity Unit Size
12345378 1 to 11 Tube 50 mm 10 Pack Each
11517392 0.5 to 5.5 Roll 5 m 4 Rolls Pack of 4
11793554 0.5 to 5 Roll 5 m 1 Roll Each
12375368 0.5 to 5 Tube 50 mm 10 Pack Each
15585669 1 to 11 Roll 5 m 1 Roll Each
15505679 1 to 14 Roll 5 m 1 Roll Each
15515679 1 to 14 Roll 5 m 4 Rolls Pack of 4
11527392 4 to 7 Roll 5 m 4 Rolls Pack of 4
12385368 5 to 9 Tube 50 mm 10 Pack Each
12395368 6.5 to 10 Tube 50 mm 10 Pack Each
11557392 9 to 13 Roll 5 m 4 Rolls Pack of 4
11713564 9 to 13 Roll 5 m 1 Roll Each
12305378 9 to 13 Tube 50 mm 10 Pack Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Product Code. 12345378
  • ONLINE EXCLUSIVE PRICE
    90.13 NOK
    77.33 NOK / Pack of 1
    Save 12.80 NOK
    14% Off
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket
Gives rough knowledge of pH.
  • Filter paper soaked with one or several colored indicators of pH
  • Available in a variety of formats to meet individual needs
  • Reference chart shows color changes in steps of pH2
Specifications
pH Range 1 to 11
Format Tube
Quantity 10 Pack
Type Grade 911
Length (Metric) 50 mm
Width (Metric) 10 mm