Learn More
Fisherbrand™ Universal Absorbent Roll
Product Code. 15843720 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Capacity (Metric):
92 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
15843720 92 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15843720
Support a Safer, More Efficient Laboratory
Use promo code 28026
View all promotions
Estimated Shipment: 19-11-2026
Add to basket
Add to basket
Fisherbrand™ Universal Absorbent Roll absorbs industrial liquids including oils, water, solvents and coolants.
  • One sided spunbound offers durability and low lint on one side
  • Multiple perforations for ease of use and to minimize waste
  • Multi-use maintenance sorbent that can be used in five different ways
    • Soc to catch leaks and drips around machines
    • Wipe to clean machines or machine parts
    • Pad to soak up small spills
    • Roll to keep the walkways safe
    • Pillow under persistent drip sources
Specifications
Color Gray
Capacity (Metric) 92 L
Material Spunbond, Meltblown
Type Universal Sorbent
For Use With (Application) Overspray, leaks and drips in drum and liquid storage areas, workstations, around and under leaky machines
Packaging Quantity 1 Roll
Thickness 2-ply
Dimensions (L x W) 46 m x 38 cm
Length (English) 150 ft.
Length (Metric) 46 m
Width (English) 14.96 in.
Width (Metric) 38 cm
Diameter (English) 2.99 in.
Diameter (Metric) 76 mm
Weight (English) 15.87 lb.
Weight (Metric) 7.2 kg