Learn More
UL27 Rabbit anti-Virus, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10363-1-AP-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16846703
  • 1415.13 NOK / 20µL
Estimated Shipment: 12-10-2023
Description
Promotes a cell cycle arrest in G0/G1 by inducing the proteasomal degradation of host histone acetyltransferase KAT5/Tip60.
Specifications
Specifications
UL27
Polyclonal
Unconjugated
UL27
UL27 Fusion Protein Ag0411
20 μL
Primary
-20°C
Liquid
ELISA
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Rabbit
Antigen Affinity Chromatography
RUO
Virus