missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UL27 Rabbit anti-Virus, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
| Antigen | UL27 |
|---|---|
| Concentration | 1 mg/mL |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16846703
|
Proteintech
10363-1-AP-20UL |
20 μL |
1710.33 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16836703
|
Proteintech
10363-1-AP-150UL |
150 μL |
4836.10 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Promotes a cell cycle arrest in G0/G1 by inducing the proteasomal degradation of host histone acetyltransferase KAT5/Tip60.Specifications
| UL27 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Virus | |
| UL27 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
| 1 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| UL27 Fusion Protein Ag0411 | |
| Primary | |
| -20°C |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title