Learn More
UCP2 293T Cell Overexpression Lysate 1 (Denatured), Abnova
Product Code. 16181722
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16181722 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16181722 Supplier Abnova Supplier No. H00007351T01.100uL
 Restricted Item
Estimated Shipment: 22-09-2026
Add to basket
Add to basket
Cell transient overexpression lysate for UCP2
Mitochondrial uncoupling proteins (UCP) are members of the larger family of mitochondrial anion carrier proteins (MACP). UCPs separate oxidative phosphorylation from ATP synthesis with energy dissipated as heat, also referred to as the mitochondrial proton leak. UCPs facilitate the transfer of anions from the inner to the outer mitochondrial membrane and the return transfer of protons from the outer to the inner mitochondrial membrane. They also reduce the mitochondrial membrane potential in mammalian cells.
  • Uncoupling protein 2 (mitochondrial, proton carrier)
  • Transfected Cell Line: 293T
  • Plasmid: pCMV-UCP2 full-length
  • Host: Human
  • Theoretical MW: 33.73kDa
  • Storage Buffer: 1X Sample Buffer (50mM Tris-HCl, 2% SDS, 10% glycerol, 300mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Western Blotting
Specifications
Cell Line 293T
For Use With (Application) Western Blot
Description UCP2 293T Cell Transient Overexpression Lysate(Denatured)
Gene Accession No. BC011737
Molecular Weight (g/mol) 33.73
pH Range 6.8
Plasmid pCMV-UCP2 full-length
Quality Control Testing Transient overexpression cell Lysate was tested with Anti-UCP2 antibody (H00007351-B01) by western blotting
Quantity 100 μL
Storage Buffer 1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Host Species Human
Content And Storage Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Show More Show Less
compliance-icons
Product Identifier
  • 100UL UCP2 293T Cell Transient Overexpression Lysa
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
  • MIXTURE LIST-Contain: Mercaptoethanol