Learn More
UBQLN2 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-23449-100UL
Description
The UBQLN2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human UBQLN2. This antibody recognizes human, rat antigen. The UBQLN2 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| UBQLN2 | |
| 1000 μg/mL | |
| CHAP1, CHAP1/DSK2, Dsk2, DSK2 homolog, hPLIC 2, HRIHFB2157, LIC 2, N4BP4, PLIC 2, PLIC2, RIHFB2157, ubiquilin 2, UBQLN2 | |
| 100 μL | |
| 29978 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
| Immunofluorescence | |
| CoraLite Plus 488 | |
| Rabbit | |
| Primary | |
| Human, Rat | |
| 150 kDa | |
| IgG |