missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TXNL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | TXNL1 |
---|---|
Concentration | 0.42 mg/mL |
Applications | Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16894404
|
Proteintech
15289-1-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16884404
|
Proteintech
15289-1-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Thioredoxins are a group of small redox active proteins possessing a conserved active site sequence. TrxL (Thioredoxin like protein 1) has 2 distinct domains: an N terminal domain, which is 43% identical to human TXN, and a C terminal domain, which shows no homology to other proteins in the sequence databases. The active site sequence, located within the N terminal domain, is that of a thioredoxin like protein; compared to the active site sequence of TXN, it has a single amino acid substitution. Unlike other thioredoxin like proteins, TrxL does not serve as a substrate for thioredoxin reductase in an insulin assay.Specifications
TXNL1 | |
Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot | |
Unconjugated | |
Rabbit | |
Mouse, Rat, Human | |
O43396, Q8CDN6, Q920J4 | |
140922, 53382, 9352 | |
TXNL1 Fusion Protein Ag7481 | |
Primary | |
-20°C | |
TXNL1 |
0.42 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
thioredoxin like 1, Thioredoxin like protein 1, TRP32, Txl, TXL 1, TXNL, TXNL1 | |
Txnl1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |