missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TXNL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15289-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Thioredoxins are a group of small redox active proteins possessing a conserved active site sequence. TrxL (Thioredoxin like protein 1) has 2 distinct domains: an N terminal domain, which is 43% identical to human TXN, and a C terminal domain, which shows no homology to other proteins in the sequence databases. The active site sequence, located within the N terminal domain, is that of a thioredoxin like protein; compared to the active site sequence of TXN, it has a single amino acid substitution. Unlike other thioredoxin like proteins, TrxL does not serve as a substrate for thioredoxin reductase in an insulin assay.Specifications
TXNL1 | |
Polyclonal | |
Unconjugated | |
TXNL1 | |
thioredoxin like 1, Thioredoxin like protein 1, TRP32, Txl, TXL 1, TXNL, TXNL1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
140922, 53382, 9352 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot | |
0.42 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O43396, Q8CDN6, Q920J4 | |
Txnl1 | |
TXNL1 Fusion Protein Ag7481 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |