Learn More
Tween 80, EssentQ, Scharlab
Product Code. 11382615
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11382615 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11382615 Supplier Scharlab Supplier No. TW00801000
  • ONLINE EXCLUSIVE PRICE
    1285.00 NOK
    1175.82 NOK / 1 L
    Save 109.18 NOK
    8% Off
 Restricted Item
Estimated Shipment: 04-01-2027