Learn More
Abnova™ TurboTEV Protease
Specifications
Specifications
| Concentration | 2 mg/mL |
| For Use With (Application) | Enzyme |
| Content And Storage | Store at -20°C, Aliquot to avoid repeated freezing and thawing |
| Form | Liquid |
| Quantity | 1 mg |
| Product Type | Protein |