Learn More
Eppendorf™ MixMate Tube Holder
Product Code. p-8006992 Shop All Eppendorf Products
Change view
Click to view available options
Tube Capacity:
0.2 mL
0.5 mL
2 mL
No. of Positions:
24
96
Unit Size:
Each
Product Code. Tube Capacity No. of Positions unitSize
10130623 0.2 mL 96 Each
10691104 0.5 mL 24 Each
10282963 2 mL 24 Each
3 options
Product Code. 10130623
Brand: Eppendorf™ 5353040113
Only null left
Add to basket
Add to basket
Designed for use with MixMate Vortex Mixer
  • Tube Holders for Eppendor MixMate™
  • The 3-in-1 mixer for plates, tubes and vortex mixing
  • High speed instrument guarantees complete mixing of your samples in seconds
  • Outstanding 2D Mix-Control provides optimized mixing with anti-spill technology
TRUSTED_SUSTAINABILITY
Specifications
Tube Capacity 0.2 mL
Product Type Tube Holder
For Use With (Equipment) 96 x 0.2mL PCR tubes, pcr strips/96-well pcr plates semi-skirted/unskirted, MixMate vortex mixer
No. of Positions 96