missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TTC12 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28216-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
TTC12 contains 3 TPR repeats. Hypermethylation of TTC12 gene may play a role in acute lymphoblastic leukemia. Haplotypic variants in DRD2, ANKK1, TTC12, and NCAM1 are associated with comorbid alcohol and drug dependence.Specifications
TTC12 | |
Polyclonal | |
Unconjugated | |
TTC12 | |
FLJ13859, FLJ20535, TPARM, TPR repeat protein 12, TTC12 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
54970 | |
-20°C | |
Liquid |
Western Blot | |
0.47 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9H892 | |
TTC12 | |
TTC12 Fusion Protein Ag28199 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |