missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TTC12 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | TTC12 |
---|---|
Concentration | 0.47 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16886055
|
Proteintech
28216-1-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16876055
|
Proteintech
28216-1-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
TTC12 contains 3 TPR repeats. Hypermethylation of TTC12 gene may play a role in acute lymphoblastic leukemia. Haplotypic variants in DRD2, ANKK1, TTC12, and NCAM1 are associated with comorbid alcohol and drug dependence.Specifications
TTC12 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q9H892 | |
54970 | |
TTC12 Fusion Protein Ag28199 | |
Primary | |
-20°C | |
TTC12 |
0.47 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
FLJ13859, FLJ20535, TPARM, TPR repeat protein 12, TTC12 | |
TTC12 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |