missing translation for 'onlineSavingsMsg'
Learn More

TTC12 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 28216-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16886055

  • 1611.84 NOK / 20µL
Estimated Shipment: 15-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

TTC12 contains 3 TPR repeats. Hypermethylation of TTC12 gene may play a role in acute lymphoblastic leukemia. Haplotypic variants in DRD2, ANKK1, TTC12, and NCAM1 are associated with comorbid alcohol and drug dependence.
Specifications

Specifications

TTC12
Polyclonal
Unconjugated
TTC12
FLJ13859, FLJ20535, TPARM, TPR repeat protein 12, TTC12
Rabbit
Antigen Affinity Chromatography
RUO
54970
-20°C
Liquid
Western Blot
0.47 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9H892
TTC12
TTC12 Fusion Protein Ag28199
20 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers