missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRPS1 Monoclonal Antibody (OTI8C11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA813180S
This item is not returnable.
View return policy
Description
TRPS1 Monoclonal antibody specifically detects TRPS1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| TRPS1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q925H1, Q9UHF7 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 299897, 7227, 83925 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI8C11 | |
| Unconjugated | |
| TRPS1 | |
| AI115454; AI447310; D15Ertd586e; GC79; LGCR; transcriptional repressor GATA binding 1; trichorhinophalangeal syndrome I; trichorhinophalangeal syndrome I (human); trichorhinophalangeal syndrome I homolog; tricho-rhino-phalangeal syndrome type I protein; TRPS1; Zinc finger protein GC79; zinc finger transcription factor Trps1 | |
| Human recombinant protein fragment corresponding to amino acids 985-1281 of human TRPS1 (NP_054831) produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction