missing translation for 'onlineSavingsMsg'
Learn More

TRPS1 Monoclonal Antibody (OTI8C11), TrueMAB™, OriGene

Product Code. p-7225525
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
Unit Size:
100µL
30µL
Product Code. Quantity unitSize
17597884 30 μL 30µL
17577884 100 μL 100µL
2 options
This item is not returnable. View return policy

Product Code. 17597884

Brand: Origene Technologies TA813180S

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TRPS1 Monoclonal antibody specifically detects TRPS1 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen TRPS1
Applications Western Blot
Classification Monoclonal
Clone OTI8C11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene TRPS1
Gene Accession No. Q925H1, Q9UHF7
Gene Alias AI115454; AI447310; D15Ertd586e; GC79; LGCR; transcriptional repressor GATA binding 1; trichorhinophalangeal syndrome I; trichorhinophalangeal syndrome I (human); trichorhinophalangeal syndrome I homolog; tricho-rhino-phalangeal syndrome type I protein; TRPS1; Zinc finger protein GC79; zinc finger transcription factor Trps1
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 985-1281 of human TRPS1 (NP_054831) produced in E.coli.
Purification Method Protein A/G
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 299897, 7227, 83925
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.