missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TrpRS Mouse anti-Human, Unconjugated, Clone: 1C6C3, Proteintech
Mouse Monoclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
| Antigen | TrpRS |
|---|---|
| Clone | 1C6C3 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17501466
|
Proteintech
67952-1-IG-20UL |
20 μL |
1710.33 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17591456
|
Proteintech
67952-1-IG-150UL |
150 μL |
4836.10 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The TrpRS antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human TrpRS. This antibody recognizes human, rat, mouse antigen. The TrpRS antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| TrpRS | |
| Western Blot | |
| Unconjugated | |
| Human, Mouse, Rat | |
| IgG2b |
| 1C6C3 | |
| Monoclonal | |
| Mouse | |
| 7453 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title