Learn More
TrpRS Mouse anti-Human, Unconjugated, Clone: 1C6C3, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67952-1-IG-20UL
 View more versions of this product
Product Code. 17501466
  • 1710.33 NOK / 20µL
Description
The TrpRS antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human TrpRS. This antibody recognizes human, rat, mouse antigen. The TrpRS antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
TrpRS
Monoclonal
Unconjugated
20 μL
7453
IgG2b
Western Blot
1C6C3
Mouse
Primary
Human, Mouse, Rat