Learn More
TrpRS Mouse anti-Human, Unconjugated, Clone: 1C6C3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67952-1-IG-20UL
Additional Details : Weight : 0.00020kg
Description
The TrpRS antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human TrpRS. This antibody recognizes human, rat, mouse antigen. The TrpRS antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
TrpRS | |
Monoclonal | |
Unconjugated | |
20 μL | |
7453 | |
IgG2b |
Western Blot | |
1C6C3 | |
Mouse | |
Primary | |
Human, Mouse, Rat |