Learn More
Miele Professional™ Trolley for washer-disinfectors
Transfer trolley MF 27/28-1 for loading and unloading washer and disinfectors
Brand: Miele Professional™ 7397640
Additional Details : Weight : 45.00000kg
Description
- Equipped with 4 castors, 2 of which can be locked
- Simple and comfortable handling thanks to rounded handles
- Dockable or lockable to rolling conveyor etc. with both sides
- Infeed height 850 mm, - 100 mm, + 150 mm
- With removable drip tray H 70, W 603, D 866 mm
Specifications
Stainless Steel | |
Transport System | |
Stainless Steel | |
Washer-disinfectors, laboratory |