missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsilylcyclopentadiene, mixture of isomers, tech. 90%

Product Code. p-7029699
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
Product Code. Quantity unitSize
11426360 1 g 1g
11436360 10 g 10g
2 options
This item is not returnable. View return policy

Product Code. 11426360

Brand: Thermo Scientific Alfa Aesar H27192.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 3559-74-8 | C8H14Si | 138.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3559-74-8
Molecular Formula C8H14Si
Molecular Weight (g/mol) 138.29
MDL Number MFCD00010666
InChI Key VMFHCJPMKUTMMQ-UHFFFAOYSA-N
Synonym 5-trimethylsilylcyclopentadiene, 5-trimethylsilyl-1,3-cyclopentadiene, 2,4-cyclopentadien-1-yl trimethylsilane, 2,4-cyclopentadien-1-yltrimethylsilane, silane, cyclopentadienyltrimethyl, 5-trimethylsilyl cyclopentadiene, trimethylsilyl cyclopentadiene, mixture of isomers, 1,3-cyclopentadiene, trimethylsilyl, silane, 2,4-cyclopentadien-1-yltrimethyl, acmc-209ii0
PubChem CID 91288
IUPAC Name cyclopenta-2,4-dien-1-yl(trimethyl)silane
SMILES C[Si](C)(C)C1C=CC=C1

Specifications

Boiling Point 138°C to 140°C
Quantity 1 g
UN Number UN1993
Flash Point 29°C (84°F)
Formula Weight 138.28
Percent Purity 90%
Grade Technical
Chemical Name or Material Trimethylsilylcyclopentadiene, mixture of isomers

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.