accessibility menu, dialog, popup

UserName

CH3SiH3CCH3

CAS RN 3559-74-8

IUPAC Name: (cyclopenta-2,4-dien-1-yl)trimethylsilane
Synonyms: cyclopenta-2,4-dien-1-yltrimethylsilane
Molecular Weight (g/mol): 138.29
Molecular Formula: C8H14Si
InChi Key: VMFHCJPMKUTMMQ-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C1C=CC=C1
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results