missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsilylcyclopentadiene, mixture of isomers, tech. 90%

Product Code. 11436360
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11436360 10 g 10g
11426360 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11436360 Supplier Thermo Scientific Alfa Aesar Supplier No. H27192.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 3559-74-8 | C8H14Si | 138.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3559-74-8
Molecular Formula C8H14Si
Molecular Weight (g/mol) 138.29
MDL Number MFCD00010666
InChI Key VMFHCJPMKUTMMQ-UHFFFAOYSA-N
Synonym 5-trimethylsilylcyclopentadiene, 5-trimethylsilyl-1,3-cyclopentadiene, 2,4-cyclopentadien-1-yl trimethylsilane, 2,4-cyclopentadien-1-yltrimethylsilane, silane, cyclopentadienyltrimethyl, 5-trimethylsilyl cyclopentadiene, trimethylsilyl cyclopentadiene, mixture of isomers, 1,3-cyclopentadiene, trimethylsilyl, silane, 2,4-cyclopentadien-1-yltrimethyl, acmc-209ii0
PubChem CID 91288
IUPAC Name cyclopenta-2,4-dien-1-yl(trimethyl)silane
SMILES C[Si](C)(C)C1C=CC=C1

Specifications

Boiling Point 138°C to 140°C
Quantity 10 g
UN Number UN1993
Flash Point 29°C (84°F)
Formula Weight 138.28
Percent Purity 90%
Grade Technical
Sensitivity Air and moisture sensitive
Density 0.833
Chemical Name or Material Trimethylsilylcyclopentadiene, mixture of isomers

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.