missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM39 Rabbit anti-Human, Mouse, HRP, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1696.32 NOK - 5307.92 NOK
Specifications
| Antigen | TRIM39 |
|---|---|
| Concentration | 1000 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | HRP |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17731489
|
Proteintech
HRP-12757-20UL |
20 μL |
1696.32 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17721489
|
Proteintech
HRP-12757-100UL |
100 μL |
5307.92 NOK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TRIM39 Polyclonal antibody specifically detects TRIM39 in Human, Mouse samples. It is validated for WBSpecifications
| TRIM39 | |
| Western Blot | |
| HRP | |
| Rabbit | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| IgG | |
| Antigen Affinity Purified | |
| Antibody |
| 1000 μg/mL | |
| Polyclonal | |
| Liquid | |
| Human, Mouse | |
| 56658 | |
| Primary | |
| Store at -20°C |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title