missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM39 Rabbit anti-Human, Mouse, HRP, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech HRP-12757-20UL
This item is not returnable.
View return policy
Description
TRIM39 Polyclonal antibody specifically detects TRIM39 in Human, Mouse samples. It is validated for WBSpecifications
| TRIM39 | |
| Polyclonal | |
| HRP | |
| NM_001786 | |
| Antigen Affinity Purified | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Western Blot | |
| 1000 μg/mL | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| Rabbit | |
| 20 μL | |
| 56658 | |
| Store at -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction