missing translation for 'onlineSavingsMsg'
Learn More

Triacetoxy(ethyl)silane, 96%

Product Code. p-7026892
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
Product Code. Quantity unitSize
11400787 100 g 100g
11410787 500 g 500g
2 options
This item is not returnable. View return policy

Product Code. 11400787

Brand: Thermo Scientific Alfa Aesar B23187.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 17689-77-9 | C8H14O6Si | 234.279 g/mol

Triacetoxy(ethyl)silane is used in sealants, the substance is used as a cross-linking agent and adhesion promoter.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triacetoxy(ethyl)silane is used in sealants, the substance is used as a cross-linking agent and adhesion promoter.

Solubility
Hydrolyzes in water.

Notes
Moisture sensitive. Store under inert gas. Store away from oxidizing agents, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17689-77-9
Molecular Formula C8H14O6Si
Molecular Weight (g/mol) 234.279
MDL Number MFCD00053703
InChI Key KXJLGCBCRCSXQF-UHFFFAOYSA-N
Synonym ethyltriacetoxysilane, triacetoxyethylsilane, triacetoxy ethyl silane, triacetoxy ethylsilane, ethylsilanetriyl triacetate, silanetriol, ethyl-, triacetate, silanetriol, 1-ethyl-, 1,1,1-triacetate, dsstox_cid_7792, dsstox_rid_78567, dsstox_gsid_27792
PubChem CID 87244
IUPAC Name [diacetyloxy(ethyl)silyl] acetate
SMILES CC[Si](OC(=O)C)(OC(=O)C)OC(=O)C

Specifications

Boiling Point 97°C to 99°C (4mmHg)
Melting Point 7°C to 9°C
Quantity 100 g
UN Number UN1760
Flash Point 106°C (224°F)
Solubility Information Hydrolyzes in water.
Formula Weight 234.28
Percent Purity 96%
Refractive Index 1.413
Sensitivity Moisture sensitive
Density 1.14
Chemical Name or Material Triacetoxy(ethyl)silane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.