CAS RN 17689-77-9
IUPAC Name:
bis(acetyloxy)(ethyl)silyl acetate
Synonyms:
bis(acetyloxy)(ethyl)silyl acetate
Molecular Weight (g/mol):
234.28
Molecular Formula:
C8H14O6Si
InChi Key:
KXJLGCBCRCSXQF-UHFFFAOYSA-N
SMILES:
CC[Si](OC(C)=O)(OC(C)=O)OC(C)=O
1
–
1
of
1
results
1
Triacetoxy(ethyl)silane, 96%
CAS: 17689-77-9 Molecular Formula: C8H14O6Si Molecular Weight (g/mol): 234.279 MDL Number: MFCD00053703 InChI Key: KXJLGCBCRCSXQF-UHFFFAOYSA-N Synonym: ethyltriacetoxysilane,triacetoxyethylsilane,triacetoxy ethyl silane,triacetoxy ethylsilane,ethylsilanetriyl triacetate,silanetriol, ethyl-, triacetate,silanetriol, 1-ethyl-, 1,1,1-triacetate,dsstox_cid_7792,dsstox_rid_78567,dsstox_gsid_27792 PubChem CID: 87244 IUPAC Name: [diacetyloxy(ethyl)silyl] acetate SMILES: CC[Si](OC(=O)C)(OC(=O)C)OC(=O)C