missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAV20 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13132-1-AP-150UL
Additional Details : Weight : 0.00010kg
Specifications
TRAV20 | |
Polyclonal | |
Unconjugated | |
TRAV20 | |
TRAV20 | |
TRAV20 Fusion Protein Ag3841 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
TCRAV20S1, TCRAV30S1, TRAV20 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
28663 | |
-20°C | |
Liquid |