missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAV20 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | TRAV20 |
---|---|
Concentration | 0.13 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16801204
|
Proteintech
13132-1-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16891194
|
Proteintech
13132-1-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
TRAV20 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
TCRAV20S1, TCRAV30S1, TRAV20 | |
TRAV20 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
28663 | |
TRAV20 Fusion Protein Ag3841 | |
Primary | |
-20°C | |
TRAV20 |