Description
- Built-in bulb eliminates the need for plugging, packaging and autoclaving
- Flexible stem bends to sample from narrow or small tubes
- Low affinity surface reduces loss of cells and valuable proteins due to binding
- Soft plastic pipets made of non-toxic low-density polyethylene (LDPE)
- Non-sterile, transparent and fully recyclable
Specifications
Specifications
| Material | LDPE |
| Drops/mL | 30 |
| Graduated | No |
| Quantity | 400 Pack |
| Tip Style | Standard |
| Volume (Metric) Dispense | 33 μL drop |
| Volume (Metric) | 1 mL |
| Length (Metric) | 104 mm |
| Type | Transfer Pipet |