Learn More
Fisherbrand™ Translucent LDPE Bottles
Product Code. p-8000414 Shop All Fisherbrand Products
Click to view available options
Capacity (Metric):
30 mL
60 mL
125 mL
250 mL
500 mL
1000 mL
Unit Size:
Pack of 12
Pack of 6
Product Code. 11547443
Only null left
Add to basket
Add to basket
Use these bottles for a variety of general storage applications. Fisherbrand™ Translucent HDPE Bottles are leak-proof and offer good chemical resistance.
  • Clear general purpose bottles
  • Blue polypropylene screw closure
Specifications
Capacity (Metric) 30 mL
Color Translucent
For Use With (Application) Ideal for general laboratory applications
Includes Polypropylene screw closure
Material Polypropylene
No. per Pack 10 Pack
Closure Color Blue
Closure Included Yes
Mouth Wide
Height (Metric) 93 mm
Product Type Wide Mouth Bottle
Closure Material Polypropylene
Closure Size 33/R3
Diameter (Metric) 36 mm
Closure Type Screw Cap