missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Transcription Termination Factor Mouse anti-Human, Mouse, Clone: 7B3H2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60235-1-IG-20UL
This item is not returnable.
View return policy
Description
The antibody is specific to TTF1.
TTF1 is a transcription factor that binds and activates the promoter of thyroid specific genes such as thyroglobulin, thyroperoxidase, and thyrotropin receptor. Crucial in the maintece of the thyroid differentiation phenotype. May play a role in lung development and surfactant homeostasis.Specifications
| Transcription Termination Factor | |
| Monoclonal | |
| 2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q15361, Q62187 | |
| TTF1 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG2a |
| Western Blot | |
| 7B3H2 | |
| Unconjugated | |
| TTF1 | |
| TTF 1, TTF I, TTF1 | |
| Mouse | |
| Protein A | |
| RUO | |
| 22130, 7270 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction