missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Transcription Termination Factor Mouse anti-Human, Mouse, Clone: 7B3H2, Proteintech
Mouse Monoclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
| Antigen | Transcription Termination Factor |
|---|---|
| Clone | 7B3H2 |
| Concentration | 2 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16847265
|
Proteintech
60235-1-IG-20UL |
20 μL |
1710.33 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16837265
|
Proteintech
60235-1-IG-150UL |
150 μL |
4836.10 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The antibody is specific to TTF1.
TTF1 is a transcription factor that binds and activates the promoter of thyroid specific genes such as thyroglobulin, thyroperoxidase, and thyrotropin receptor. Crucial in the maintece of the thyroid differentiation phenotype. May play a role in lung development and surfactant homeostasis.Specifications
| Transcription Termination Factor | |
| 2 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| TTF 1, TTF I, TTF1 | |
| TTF1 | |
| IgG2a | |
| Protein A | |
| Antibody |
| 7B3H2 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Mouse, Human | |
| Q15361, Q62187 | |
| 22130, 7270 | |
| Peptide | |
| Primary | |
| -20°C | |
| TTF1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title