missing translation for 'onlineSavingsMsg'
Learn More

trans,trans-4-(3,4-Difluorophenyl)-4'-n-pentylbicyclohexyl, 97%

Product Code. 15461946
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15461946 5 g 5g
15451946 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15461946 Supplier Thermo Scientific Alfa Aesar Supplier No. H55168.06
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 118164-51-5 | C23H34F2 | 348.522 g/mol

It is used as flavor and fragrance intermediate. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as flavor and fragrance intermediate. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

Solubility
Slightly soluble in water.

Notes
Reacts with strong oxidizing agents. Keep at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118164-51-5
Molecular Formula C23H34F2
Molecular Weight (g/mol) 348.522
MDL Number MFCD13182310
InChI Key YDVUSMRUBCJGAV-UHFFFAOYSA-N
Synonym trans,trans-4-3,4-difluorophenyl-4'-pentyl-1,1'-bi cyclohexane, trans,trans-4-3,4-difluorophenyl-4'-pentylbicyclohexyl, 4-trans-4-trans-4-pentylcyclohexyl cyclohexyl-1,2-difluorobenzene, benzene, 1,2-difluoro-4-4'-pentyl 1,1'-bicyclohexyl-4-yl, 1,2-difluoro-4-trans,trans-4'-pentyl 1,1'-bicyclohexyl-4-yl benzene, 1,2-difluoro-4-4-4-pentylcyclohexyl cyclohexyl benzene, 4-3,4-difluorophenyl-4'-pentyl-1,1'-bi cyclohexane, trans,trans-4-3,4-difluorophenyl-4'-n-pentylbicyclohexyl, 1r,1's,4r,4'r-4-3,4-difluorophenyl-4'-pentyl-1,1'-bi cyclohexane, trans,trans-4-pentylbicyclohexyl-3,4-difluorobenzene
PubChem CID 581521
IUPAC Name 1,2-difluoro-4-[4-(4-pentylcyclohexyl)cyclohexyl]benzene
SMILES CCCCCC1CCC(CC1)C2CCC(CC2)C3=CC(=C(C=C3)F)F

Specifications

Melting Point 45°C to 47°C
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 348.52
Percent Purity 97%
Chemical Name or Material trans,trans-4-(3,4-Difluorophenyl)-4'-n-pentylbicyclohexyl

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.