accessibility menu, dialog, popup

UserName

H3CFF

CAS RN 118164-51-5

IUPAC Name: 4-(3,4-difluorophenyl)-4'-pentyl-1,1'-bi(cyclohexane)
Synonyms: 4-(3,4-difluorophenyl)-4'-pentyl-1,1'-bi(cyclohexane)
Molecular Weight (g/mol): 348.52
Molecular Formula: C23H34F2
InChi Key: YDVUSMRUBCJGAV-UHFFFAOYSA-N
SMILES: CCCCCC1CCC(CC1)C1CCC(CC1)C1=CC(F)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results