missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Benzyl-L-proline hydrochloride, 95%, Thermo Scientific™

Product Code. 15478527
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15478527 250 mg 250mg
15468527 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15478527 Supplier Thermo Scientific Alfa Aesar Supplier No. H52041.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1279049-67-0
Molecular Formula C12H16ClNO2
Molecular Weight (g/mol) 241.715
MDL Number MFCD15143631
InChI Key GPWGPOYNAWMDDN-DHXVBOOMSA-N
Synonym 2s,4r-4-benzylpyrrolidine-2-carboxylic acid hydrochloride, trans-4-benzyl-l-proline hydrochloride, r-gamma-benzyl-l-proline-hcl, r-, a-benzyl-l-proline inverted exclamation mark currencyhcl, 4r-4-benzyl-l-proline hydrochloride, 2s,4r-4-benzylpyrrolidine-2-carboxylic acid hcl, 2s,4r-4-benzylpyrrolidine-2-carboxylicacidhydrochloride
PubChem CID 2761962
IUPAC Name (2S,4R)-4-benzylpyrrolidine-2-carboxylic acid;hydrochloride
SMILES C1C(CNC1C(=O)O)CC2=CC=CC=C2.Cl

Specifications

Quantity 250 mg
Formula Weight 241.72
Percent Purity 95%
Chemical Name or Material trans-4-Benzyl-L-proline hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.