accessibility menu, dialog, popup

UserName

CAS RN 1279049-67-0

IUPAC Name: 4-benzylpyrrolidine-2-carboxylic acid hydrochloride
Synonyms: 4-benzylpyrrolidine-2-carboxylic acid hydrochloride
Molecular Weight (g/mol): 241.72
Molecular Formula: C12H16ClNO2
InChi Key: GPWGPOYNAWMDDN-UHFFFAOYNA-N
SMILES: Cl.OC(=O)C1CC(CC2=CC=CC=C2)CN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results