Learn More
trans-4-Aminocyclohexanecarboxylic acid hydrochloride, 96%, Thermo Scientific Chemicals
Product Code. 15427795
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15427795 5 g 5g
15417795 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15427795 Supplier Thermo Scientific Alfa Aesar Supplier No. H31114.06
Request Bulk or Custom Format
Used as intermediate in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used as intermediate in organic synthesis.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 27960-59-4
Molecular Formula C7H14ClNO2
Molecular Weight (g/mol) 179.644
MDL Number MFCD03844587
InChI Key HXZSYUOXTKQNNY-UHFFFAOYSA-N
Synonym trans-4-aminocyclohexanecarboxylic acid hydrochloride, cis-4-aminocyclohexanecarboxylic acid hydrochloride, 4-aminocyclohexanecarboxylic acid hydrochloride, 4-aminocyclohexane-1-carboxylic acid hydrochloride, trans-4-aminocyclohexane carboxylic acid hydrochloride, trans-4-aminocyclohexanecarboxylic acid hcl, trans-4-aminocyclohexane-1-carboxylic acid hydrochloride, trans-4-amino-cyclohexane carboxylic acid hydrochloride, 1r,4r-4-aminocyclohexane-1-carboxylic acid hydrochloride, cis-4-aminocyclohexanecarboxylic acid hcl
PubChem CID 24720880
IUPAC Name 4-aminocyclohexane-1-carboxylic acid;hydrochloride
SMILES C1CC(CCC1C(=O)O)N.Cl
Specifications
Melting Point >300°C
Solubility Information Soluble in water.
Quantity 5 g
Formula Weight 179.65
Percent Purity 96%
Chemical Name or Material trans-4-Aminocyclohexanecarboxylic acid hydrochloride