accessibility menu, dialog, popup

UserName

CAS RN 27960-59-4

OHONH2HCl

CAS RN 27960-59-4

IUPAC Name: 4-aminocyclohexane-1-carboxylic acid hydrochloride
Synonyms: 4-aminocyclohexane-1-carboxylic acid hydrochloride
Molecular Weight (g/mol): 179.64
Molecular Formula: C7H14ClNO2
InChi Key: HXZSYUOXTKQNNY-UHFFFAOYSA-N
SMILES: Cl.NC1CCC(CC1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)