missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Aminocyclohexanecarboxylic acid hydrochloride, 96%, Thermo Scientific Chemicals

Product Code. 15417795
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15417795 1 g 1g
15427795 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15417795 Supplier Thermo Scientific Alfa Aesar Supplier No. H31114.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Used as intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as intermediate in organic synthesis.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27960-59-4
Molecular Formula C7H14ClNO2
Molecular Weight (g/mol) 179.644
MDL Number MFCD03844587
InChI Key HXZSYUOXTKQNNY-UHFFFAOYSA-N
Synonym trans-4-aminocyclohexanecarboxylic acid hydrochloride, cis-4-aminocyclohexanecarboxylic acid hydrochloride, 4-aminocyclohexanecarboxylic acid hydrochloride, 4-aminocyclohexane-1-carboxylic acid hydrochloride, trans-4-aminocyclohexane carboxylic acid hydrochloride, trans-4-aminocyclohexanecarboxylic acid hcl, trans-4-aminocyclohexane-1-carboxylic acid hydrochloride, trans-4-amino-cyclohexane carboxylic acid hydrochloride, 1r,4r-4-aminocyclohexane-1-carboxylic acid hydrochloride, cis-4-aminocyclohexanecarboxylic acid hcl
PubChem CID 24720880
IUPAC Name 4-aminocyclohexane-1-carboxylic acid;hydrochloride
SMILES C1CC(CCC1C(=O)O)N.Cl

Specifications

Melting Point >300°C
Solubility Information Soluble in water.
Quantity 1 g
Formula Weight 179.65
Percent Purity 96%
Chemical Name or Material trans-4-Aminocyclohexanecarboxylic acid hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.