Learn More
Fisherbrand™ Traceable™ Data Logging Ethernet Thermometers Compatible with TraceableLIVE™ Cloud Service
Product Code. 16536820 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Temperature Range (Metric):
-50°C to +60°C
-90°C to +105°C
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature Range (Metric) Unit Size
16536820 -90°C to +105°C Each
16526820 -50°C to +60°C Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16536820
Estimated Shipment: 13-05-2027
Add to basket
Add to basket
Thermometer that allows you to stay connected to your critical environments no matter where you go.
  • Connect to your data via smartphone, tablet, or PC using TraceableLIVE cloud service
  • Get the reliability, speed, and security using Ethernet connection
  • View current temperature conditions anywhere – cloud-based interface sends email, text, and push notifications
  • No software required – web, Android™, Apple™ iOS data visibility
  • Get accurate, reliable measurements with included NIST-traceable calibration
Specifications
Type Thermometer
Calibration Calibrated
Includes One stainless steel probe with 10 ft. (3 m) long cable, holder with magnetic back, Ethernet cable, universal power adapter and plug with 5 ft. (1.5 m) long cord, three AAA alkaline batteries, and NIST-traceable calibration certificate with data
Material Plastic
For Use With (Application) Temperature monitoring
Temperature Range (Metric) -90°C to +105°C
Length (Metric) 6.985 cm
Weight (Metric) 0.544 kg