Learn More
Amarell™ Total Immersion Laboratory Thermometers
Product Code. 10549481
Change view
Click to view available options
Temperature Range (Metric):
-50°C to +50°C
Subdivision:
1°C
5°C
Length (Metric):
300 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature Range (Metric) Subdivision Length (Metric) unitSize
10549481 -50°C to +50°C 1°C 300 mm Each
10052573 -50°C to +50°C 5°C 300 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10549481 Supplier Amarell™ Supplier No. L33004
Specifications
Certifications/Compliance DIN 12 778
Immersion Type Total Immersion
Length (Metric) 300 mm
Thermometer Fill Toluene
Product Type Thermometer
Max. Temperature (Metric) 50°C
Min. Temperature (Metric) -50°C
Temperature Range (Metric) -50°C to +50°C
Calibrated Yes
Subdivision 1°C