Learn More
Unity Lab Services™ Total Care Warranty for Thermo Scientific Bio Safety Cabinets
Product Code. 17260394
Change view
Click to view available options
For Use With (Equipment):
Biosafety Cabinet (0.9 to 1.5 m)
Biosafety Cabinet (1.8 m)
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
17260394 Biosafety Cabinet (0.9 to 1.5 m) Each
17270394 Biosafety Cabinet (1.8 m) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17260394 Supplier Unity Lab Services™ Supplier No. TCWTYBSC1
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
 Restricted Item
Estimated Shipment: 03-02-2027
Add to basket
Add to basket
Total Care Warranty provides an upgrade to factory warranty. Includes 2 day response time (within specified location and criteria), priority technical support, and one preventative maintenance visit.
  • Upgrade a basic factory warranty with priority Technical Support response, and faster on-site scheduling along with an annual preventative maintenance visit
  • 2 business day response time (may vary by location) for corrective maintenance
  • Includes parts and labor covered under factory warranty for corrective maintenance
  • One scheduled Preventative Maintenance visit, including a preventative maintenance kit (where applicable)
  • Priority unlimited access to OEM-trained Technical support experts who can diagnose, investigate, and remotely resolve issues or schedule on-site visits using the latest digital remote support tools.
  • Access to online service knowledge base
  • Corrective Maintenance Repair Guarantee for the duration of product warranty or 1 year whichever is longer, as defined in the terms and conditions
  • Instrument Software/ Firmware updates included as needed to support a preventative maintenance or corrective maintenance visit
  • Upgrades to new versions of software are not included
  • 10% discount on additional services including additional preventative maintenance visits, calibration, and qualification services
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Biosafety Cabinet (0.9 to 1.5 m)
For Use With (Application) Upgrade to Factory Warranty
Warranty and Service Offering Priority Technical Support, and One Preventative Maintenance Visit
Includes 2 Day Response Time (Within Specified Location and Criteria), Priority Technical Support, and One Preventative Maintenance Visit
Product Type Total Care Warranty
Warranty Response Time 2 Days (Within Specified Location and Criteria)
Warranty Services Not Included Upgrades to New Software Versions