Learn More
Unity Lab Services™ Total Care Service Plan for Thermo Scientific Bio Safety Cabinets
Product Code. 17230434
Change view
Click to view available options
For Use With (Equipment):
Biosafety Cabinet (0.9 to 1.5 m)
Biosafety Cabinet (1.8 m)
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
17230434 Biosafety Cabinet (0.9 to 1.5 m) Each
17240434 Biosafety Cabinet (1.8 m) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17230434 Supplier Unity Lab Services™ Supplier No. TLCBSC1
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket
Total Care Service Plan provides support post factory warranty. Includes 2 day response time (within specified location and criteria), full parts and labor coverage, and one preventative maintenance visit.
  • Continue comprehensive service support post factory warranty with faster response time, and proactive maintenance through annual preventative maintenance visit (or event)
  • 2 business day response time (may vary by location) for corrective maintenance
  • Includes Engineer labor, travel, and factory-certified replacement parts required for corrective maintenance
  • One scheduled Preventative Maintenance visit including a preventative maintenance kit (where applicable)
  • Priority unlimited access to OEM-trained Technical support experts who can diagnose, investigate, and remotely resolve issues or schedule on-site visits using the latest digital remote support tools
  • Access to on-line service knowledge base
  • Corrective Maintenance Repair Guarantee for the duration of product warranty or 1 year whichever is longer, as defined in the contract terms and conditions
  • Instrument Software/ Firmware updates included as needed to support a preventative maintenance or corrective maintenance visit
  • Upgrades to new versions of software are not included
  • 10% discount on additional services including additional preventative maintenance visits, calibration, and qualification services
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Biosafety Cabinet (0.9 to 1.5 m)
For Use With (Application) Support Post Factory Warranty
Includes 2 Day Response Time (Within Specified Location and Criteria), Full Parts & Labor Coverage, and One Preventative Maintenance Visit
Product Type Total Care Service Plan
Services Not Included Upgrades to New Software Versions
Warranty Response Time 2 Days (Within Specified Location and Criteria)
Services Included Full Parts & Labor Coverage, and One Preventative Maintenance Visit