missing translation for 'onlineSavingsMsg'
Learn More

Fesoterodine-d7, TRC

Product Code. 30179772
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179772 5 mg 5mg
30180395 25 mg 25mg
30180196 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30179772 Supplier Toronto Research Chemicals Supplier No. TRCF3213035MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1126611-84-4
Molecular Formula C26H30D7NO3
Molecular Weight (g/mol) 418.62
SMILES [2H]C([2H])([2H])C([2H])(C(=O)Oc1ccc(CO)cc1[C@H](CCN(C(C)C)C(C)C)c2ccccc2)C([2H])([2H])[2H]

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C26H37NO3/c1-18(2)26(29)30-25-13-12-21(17-28)16-24(25)23(22-10-8-7-9-11-22)14-15-27(19(3)4)20(5)6/h7-13,16,18-20,23,28H,14-15,17H2,1-6H3/t23-/m1/s1/i1D3,2D3,18D
Formula Weight 418.3213
Chemical Name or Material Fesoterodine-d7
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.