CAS RN 1126611-84-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Fesoterodine-d7, TRC
CAS: 1126611-84-4 Molecular Formula: C26H30D7NO3 Molecular Weight (g/mol): 418.62 SMILES: [2H]C([2H])([2H])C([2H])(C(=O)Oc1ccc(CO)cc1[C@H](CCN(C(C)C)C(C)C)c2ccccc2)C([2H])([2H])[2H]